tert-Butyl-4-[2-(4-bromo-1H-pyrazol-1-yl)ethyl]piperazine-1-carboxylate ≥97%
Supplier: Matrix Scientific
083922-1G
102841-084EA
636.92
USD
102841-084
tert-Butyl-4-[2-(4-bromo-1H-pyrazol-1-yl)ethyl]piperazine-1-carboxylate ≥97%
tert-Butyl-4-[2-(4-bromo-1H-pyrazol-1-yl)ethyl]piperazine-1-carboxylate
Hazard: Irritant
Formula:
C₁₄H₂₃BrN₄O₂ MW: 359.27 g/mol |
MDL Number:
MFCD11100120 CAS Number: 877401-28-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...