1-Benzyl-4-(2-iodo-1H-pyrrol-1-yl)piperidine ≥97%

Supplier: Matrix Scientific

083909-1G
102841-046EA 707.69 USD
102841-046
1-Benzyl-4-(2-iodo-1H-pyrrol-1-yl)piperidine ≥97%
1-Benzyl-4-(2-iodo-1H-pyrrol-1-yl)piperidine

Hazard: Irritant

Formula: C₁₆H₁₉IN₂
MDL Number: MFCD09864949
CAS Number: 254115-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR