5-(5-Bromo-2-furyl)cyclohexane-1,3-dione ≥97%
Supplier: Matrix Scientific
083908-1G
102841-044EA
863.08
USD
102841-044
5-(5-Bromo-2-furyl)cyclohexane-1,3-dione ≥97%
5-(5-Bromo-2-furyl)cyclohexane-1,3-dione
Hazard: Irritant
Formula:
C₁₀H₉BrO₃ |
MDL Number:
MFCD12827891 CAS Number: 1216614-19-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...