[5-(5-Bromo-2-furyl)-2-oxo-1,3,4-oxadiazol-3(2H)-yl]acetic acid ≥97%
Supplier: Matrix Scientific
083906-1G
102841-040EA
863.08
USD
102841-040
[5-(5-Bromo-2-furyl)-2-oxo-1,3,4-oxadiazol-3(2H)-yl]acetic acid ≥97%
[5-(5-Bromo-2-furyl)-2-oxo-1,3,4-oxadiazol-3(2H)-yl]acetic acid
Hazard: Irritant
Formula:
C₈H₅BrN₂O₅ |
MDL Number:
MFCD09991802 CAS Number: 1086376-35-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...