5-(4-Bromo-3-methoxythien-2-yl)-1,3,4-oxadiazol-2(3H)-one ≥97%

Supplier: Matrix Scientific

083899-1G
102841-020EA 693.85 USD
102841-020
5-(4-Bromo-3-methoxythien-2-yl)-1,3,4-oxadiazol-2(3H)-one ≥97%
5-(4-Bromo-3-methoxythien-2-yl)-1,3,4-oxadiazol-2(3H)-one

Hazard: Irritant

Formula: C₇H₅BrN₂O₃S
MDL Number: MFCD09991801
CAS Number: 1086376-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR