3-Chloro-2-(cyclohexyloxy)-5-(trifluoromethyl)pyridine ≥97%
Supplier: Matrix Scientific
083851-1G
102840-900EA
636.92
USD
102840-900
3-Chloro-2-(cyclohexyloxy)-5-(trifluoromethyl)pyridine ≥97%
3-Chloro-2-(cyclohexyloxy)-5-(trifluoromethyl)pyridine
Hazard: Irritant
Formula:
C₁₂H₁₃ClF₃NO |
MDL Number:
MFCD11840288 CAS Number: 1216838-34-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...