3-Chloro-2-(tetrahydro-2H-pyran-4-yloxy)-5-(trifluoromethyl)pyridine ≥97%

Supplier: Matrix Scientific

083850-1G
102840-898EA 666.15 USD
102840-898
3-Chloro-2-(tetrahydro-2H-pyran-4-yloxy)-5-(trifluoromethyl)pyridine ≥97%
3-Chloro-2-(tetrahydro-2H-pyran-4-yloxy)-5-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₁₁H₁₁ClF₃NO₂
MDL Number: MFCD11840287
CAS Number: 1217013-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR