3{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]thio}propanohydrazide ≥97%

Supplier: Matrix Scientific

083845-1G
102840-888EA 553.85 USD
102840-888
3{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]thio}propanohydrazide ≥97%
3{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]thio}propanohydrazide

Hazard: Irritant

Formula: C₉H₉ClF₃N₃OS
MW: 299.7 g/mol
MDL Number: MFCD11100119
CAS Number: 1215738-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR