Methyl-3-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]thio}propanoate ≥97%

Supplier: Matrix Scientific

083844-1G
102840-886EA 524.62 USD
102840-886
Methyl-3-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]thio}propanoate ≥97%
Methyl-3-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]thio}propanoate

Hazard: Irritant

Formula: C₁₀H₉ClF₃NO₂S
MW: 299.7 g/mol
MDL Number: MFCD03153507
CAS Number: 380453-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR