2-Chloro-6-piperidin-1-yl-4-(trifluoromethyl)pyridine ≥97%
Supplier: Matrix Scientific
083836-1G
102840-868EA
636.92
USD
102840-868
2-Chloro-6-piperidin-1-yl-4-(trifluoromethyl)pyridine ≥97%
2-Chloro-6-piperidin-1-yl-4-(trifluoromethyl)pyridine
Hazard: Irritant
Formula:
C₁₁H₁₂ClF₃N₂ |
MDL Number:
MFCD09991800 CAS Number: 1086376-19-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...