{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]amino}acetic acid ≥97%

Supplier: Matrix Scientific

083834-1G
102840-860EA 566.15 USD
102840-860
{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]amino}acetic acid ≥97%
{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]amino}acetic acid

Hazard: Irritant

Formula: C₈H₆ClF₃N₂O₂
MDL Number: MFCD09864947
CAS Number: 1016690-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR