1-Bromo-3-[(phenylsulfonyl)methyl]benzene ≥97%

Supplier: Matrix Scientific

083821-1G
102840-822EA 636.92 USD
102840-822
1-Bromo-3-[(phenylsulfonyl)methyl]benzene ≥97%
1-Bromo-3-[(phenylsulfonyl)methyl]benzene

Hazard: Irritant

Formula: C₁₃H₁₁BrO₂S
MDL Number: MFCD11840285
CAS Number: 130891-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR