2-(3-Bromophenoxy)-3-chloro-5-(trifluoromethyl)pyridine ≥97%

Supplier: Matrix Scientific

083819-1G
102840-818EA 383.08 USD
102840-818
2-(3-Bromophenoxy)-3-chloro-5-(trifluoromethyl)pyridine ≥97%
2-(3-Bromophenoxy)-3-chloro-5-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₁₂H₆BrClF₃NO
MDL Number: MFCD11840283
CAS Number: 1041591-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR