4-(3-Bromophenoxy)-2-chloropyrimidine ≥97%

Supplier: Matrix Scientific

083816-1G
102840-812EA 467.69 USD
102840-812
4-(3-Bromophenoxy)-2-chloropyrimidine ≥97%
4-(3-Bromophenoxy)-2-chloropyrimidine

Hazard: Irritant

Formula: C₁₀H₆BrClN₂O
MDL Number: MFCD11505038
CAS Number: 956039-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR