1-(3-Bromobenzyl)-1H-1,2,4-triazole ≥97%
Supplier: Matrix Scientific
083804-1G
102840-776EA
636.92
USD
102840-776
1-(3-Bromobenzyl)-1H-1,2,4-triazole ≥97%
1-(3-Bromobenzyl)-1H-1,2,4-triazole
Hazard: Irritant
Formula:
C₉H₈BrN₃ |
MDL Number:
MFCD11100117 CAS Number: 942154-19-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...