5-[(E)-2-(2-Chloro-3,4-dimethoxyphenyl)vinyl]-1,3,4-oxadiazol-2(3H)-one ≥97%
Supplier: Matrix Scientific
083794-1G
102840-748EA
340
USD
102840-748
5-[(E)-2-(2-Chloro-3,4-dimethoxyphenyl)vinyl]-1,3,4-oxadiazol-2(3H)-one ≥97%
5-[(E)-2-(2-Chloro-3,4-dimethoxyphenyl)vinyl]-1,3,4-oxadiazol-2(3H)-one
Hazard: Irritant
Formula:
C₁₂H₁₁ClN₂O₄ MW: 282.69 g/mol |
MDL Number:
MFCD09991796 CAS Number: 1086376-13-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...