1-(3,4-Difluorobenzyl)-1H-pyrrole ≥97%

Supplier: Matrix Scientific

083759-1G
102840-650EA 636.92 USD
102840-650
1-(3,4-Difluorobenzyl)-1H-pyrrole ≥97%
1-(3,4-Difluorobenzyl)-1H-pyrrole

Hazard: Irritant

Formula: C₁₁H₉F₂N
MDL Number: MFCD14684087
CAS Number: 1247669-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR