1-Iodo-2-(methylsulfinyl)benzene ≥97%

Supplier: Matrix Scientific

083749-1G
102840-624EA 920 USD
102840-624
1-Iodo-2-(methylsulfinyl)benzene ≥97%
1-Iodo-2-(methylsulfinyl)benzene

Hazard: Irritant

Formula: C₇H₇IOS
MW: 266.1 g/mol
MDL Number: MFCD18089236
CAS Number: 71545-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR