Ethyl-3-amino-6-methyl-4-(trifluoromethyl)thieno[2,3-b]pyridine-2-carboxylate ≥97%

Supplier: Matrix Scientific

083669-1G
102840-362EA 750.77 USD
102840-362
Ethyl-3-amino-6-methyl-4-(trifluoromethyl)thieno[2,3-b]pyridine-2-carboxylate ≥97%
Ethyl-3-amino-6-methyl-4-(trifluoromethyl)thieno[2,3-b]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₁F₃N₂O₂S
MDL Number: MFCD00486827
CAS Number: 299198-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR