Methyl-3-chloroimidazo[1,2-a]pyridine-6-carboxylate ≥97%

Supplier: Matrix Scientific

083650-1G
102840-320EA 566.15 USD
102840-320
Methyl-3-chloroimidazo[1,2-a]pyridine-6-carboxylate ≥97%
Methyl 3-chloroimidazo[1,2-a]pyridine-6-carboxylate

Hazard: Irritant

Formula: C₉H₇ClN₂O₂
MW: 210.62
MDL Number: MFCD07021500
CAS Number: 900019-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR