Ethyl-2-[(4-fluorobenzyl)amino]cyclopentanecarboxylate ≥97%
Supplier: Matrix Scientific
083626-1G
102840-266EA
806.15
USD
102840-266
Ethyl-2-[(4-fluorobenzyl)amino]cyclopentanecarboxylate ≥97%
Ethyl-2-[(4-fluorobenzyl)amino]cyclopentanecarboxylate
Hazard: Irritant
Formula:
C₁₅H₂₀FNO₂ |
MDL Number:
MFCD19706057 CAS Number: 1355334-68-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...