Ethyl-2-(cyclohexylamino)cyclopentanecarboxylate ≥97%

Supplier: Matrix Scientific

083624-1G
102840-262EA 806.15 USD
102840-262
Ethyl-2-(cyclohexylamino)cyclopentanecarboxylate ≥97%
Ethyl-2-(cyclohexylamino)cyclopentanecarboxylate

Hazard: Irritant

Formula: C₁₄H₂₅NO₂
MDL Number: MFCD19706053
CAS Number: 1355334-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR