Ethyl-1-(4-nitrobenzoyl)cyclopropanecarboxylate ≥97%

083616-1G
102840-236EA 497.95 USD
102840-236
Ethyl-1-(4-nitrobenzoyl)cyclopropanecarboxylate ≥97%
Ethyl-1-(4-nitrobenzoyl)cyclopropanecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₃NO₅
MDL Number: MFCD16622251
CAS Number: 1287217-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR