Ethyl-1-benzoylcyclopropanecarboxylate ≥97%

083615-1G
102840-234EA 645.15 USD
102840-234
Ethyl-1-benzoylcyclopropanecarboxylate ≥97%
Ethyl-1-benzoylcyclopropanecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₄O₃
MDL Number: MFCD16622250
CAS Number: 116195-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR