tert-Butyl (1R)-1-(tert-butylamino)-4-oxocyclopent-2-ene-1-carboxylate ≥97%

Supplier: Matrix Scientific

083614-1G
102840-232EA 1118.46 USD
102840-232
tert-Butyl (1R)-1-(tert-butylamino)-4-oxocyclopent-2-ene-1-carboxylate ≥97%
tert-Butyl (1R)-1-(tert-butylamino)-4-oxocyclopent-2-ene-1-carboxylate

Hazard: Irritant

Formula: C₁₄H₂₃NO₃
MDL Number: MFCD13195610
CAS Number: 1235439-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR