Methyl-1-cyclohexyl-5-oxopyrrolidine-3-carboxylate ≥97%

Supplier: Matrix Scientific

083592-1G
102840-168EA 750.77 USD
102840-168
Methyl-1-cyclohexyl-5-oxopyrrolidine-3-carboxylate ≥97%
Methyl-1-cyclohexyl-5-oxopyrrolidine-3-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₉NO₃
MDL Number: MFCD00490451
CAS Number: 100252-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR