Methyl-5-oxo-1-phenylpyrrolidine-2-carboxylate ≥97%

Supplier: Matrix Scientific

083589-1G
102840-162EA 806.15 USD
102840-162
Methyl-5-oxo-1-phenylpyrrolidine-2-carboxylate ≥97%
Methyl-5-oxo-1-phenylpyrrolidine-2-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₃NO₃
MDL Number: MFCD10004229
CAS Number: 146500-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR