Methyl-5-oxo-1-piperidin-3-ylpyrrolidine-3-carboxylate ≥97%

Supplier: Matrix Scientific

083588-1G
102840-160EA 636.92 USD
102840-160
Methyl-5-oxo-1-piperidin-3-ylpyrrolidine-3-carboxylate ≥97%
Methyl-5-oxo-1-piperidin-3-ylpyrrolidine-3-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₈N₂O₃
MDL Number: MFCD14666499
CAS Number: 1287218-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR