Methyl-1-phenyl-5-thioxopyrrolidine-3-carboxylate ≥97%
Supplier: Matrix Scientific
083585-1G
102840-154EA
467.69
USD
102840-154
Methyl-1-phenyl-5-thioxopyrrolidine-3-carboxylate ≥97%
Methyl-1-phenyl-5-thioxopyrrolidine-3-carboxylate
Hazard: Irritant
Formula:
C₁₂H₁₃NO₂S |
MDL Number:
MFCD10703537 CAS Number: 1086375-83-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...