Ethyl-1-(3-aminobenzyl)-5-methyl-1H-1,2,3-triazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

083553-1G
102840-070EA 636.92 USD
102840-070
Ethyl-1-(3-aminobenzyl)-5-methyl-1H-1,2,3-triazole-4-carboxylate ≥95%
Ethyl-1-(3-aminobenzyl)-5-methyl-1H-1,2,3-triazole-4-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₆N₄O₂
MDL Number: MFCD09864942
CAS Number: 1086375-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR