Ethyl-2-nitrilo-2-(2-oxoindolin-3-ylidene)acetate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-cyano-2-(2-oxo-1,2-dihydro-3H-indol-3-yliden)acetate

083535-1G
102840-030EA 666.15 USD
102840-030
Ethyl-2-nitrilo-2-(2-oxoindolin-3-ylidene)acetate ≥95%
Ethyl-2-nitrilo-2-(2-oxoindolin-3-ylidene)acetate

Hazard: Irritant

Formula: C₁₃H₁₀N₂O₃
MW: 242.24 g/mol
MDL Number: MFCD00170750
CAS Number: 14003-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR