Ethyl-3-amino-5-(trifluoromethyl)-1-benzothiophene-2-carboxylate ≥97%
Supplier: Matrix Scientific
083531-1G
102840-020EA
403.08
USD
102840-020
Ethyl-3-amino-5-(trifluoromethyl)-1-benzothiophene-2-carboxylate ≥97%
Ethyl-3-amino-5-(trifluoromethyl)-1-benzothiophene-2-carboxylate
Hazard: Irritant
Formula:
C₁₂H₁₀F₃NO₂S MW: 289.28 g/mol |
MDL Number:
MFCD11505029 CAS Number: 1217018-83-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...