Ethyl-3-amino-6-bromo-1-benzothiophene-2-carboxylate ≥97%

Supplier: Matrix Scientific

083530-1G
102840-018EA 383.08 USD
102840-018
Ethyl-3-amino-6-bromo-1-benzothiophene-2-carboxylate ≥97%
Ethyl-3-amino-6-bromo-1-benzothiophene-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₀BrNO₂S
MW: 300.18 g/mol
MDL Number: MFCD11505028
CAS Number: 1216708-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR