Ethyl-3-amino-4-iodo-1-benzothiophene-2-carboxylate ≥97%
Supplier: Matrix Scientific
083529-1G
102840-016EA
750.77
USD
102840-016
Ethyl-3-amino-4-iodo-1-benzothiophene-2-carboxylate ≥97%
Ethyl-3-amino-4-iodo-1-benzothiophene-2-carboxylate
Hazard: Irritant
Formula:
C₁₁H₁₀INO₂S MW: 347.18 g/mol |
MDL Number:
MFCD11505027 CAS Number: 1216596-20-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...