Ethyl-5-(3-methoxybenzyl)-1,2,4-oxadiazole-3-carboxylate ≥97%
Supplier: Matrix Scientific
083523-1G
102840-000EA
636.92
USD
102840-000
Ethyl-5-(3-methoxybenzyl)-1,2,4-oxadiazole-3-carboxylate ≥97%
Ethyl-5-(3-methoxybenzyl)-1,2,4-oxadiazole-3-carboxylate
Hazard: Irritant
Formula:
C₁₃H₁₄N₂O₄ |
MDL Number:
MFCD11100108 CAS Number: 1216607-98-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...