Ethyl-5-(4-bromobenzyl)-1,2,4-oxadiazole-3-carboxylate ≥97%

Supplier: Matrix Scientific

083522-1G
102839-998EA 636.92 USD
102839-998
Ethyl-5-(4-bromobenzyl)-1,2,4-oxadiazole-3-carboxylate ≥97%
Ethyl-5-(4-bromobenzyl)-1,2,4-oxadiazole-3-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₁BrN₂O₃
MDL Number: MFCD11100107
CAS Number: 1216958-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR