Ethyl-5-(2-bromobenzyl)-1,2,4-oxadiazole-3-carboxylate ≥97%
Supplier: Matrix Scientific
083521-1G
102839-996EA
636.92
USD
102839-996
Ethyl-5-(2-bromobenzyl)-1,2,4-oxadiazole-3-carboxylate ≥97%
Ethyl-5-(2-bromobenzyl)-1,2,4-oxadiazole-3-carboxylate
Hazard: Irritant
Formula:
C₁₂H₁₁BrN₂O₃ |
MDL Number:
MFCD11100106 CAS Number: 1216696-40-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...