Ethyl-5-(3-furyl)-1,2,4-oxadiazole-3-carboxylate ≥97%

Supplier: Matrix Scientific

083519-1G
102839-992EA 806.15 USD
102839-992
Ethyl-5-(3-furyl)-1,2,4-oxadiazole-3-carboxylate ≥97%
Ethyl-5-(3-furyl)-1,2,4-oxadiazole-3-carboxylate

Hazard: Irritant

Formula: C₉H₈N₂O₄
MDL Number: MFCD09991791
CAS Number: 1086375-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR