tert-Butyl-6-oxo-1,6-dihydropyridazine-3-carboxylate ≥97%

Supplier: Matrix Scientific

083512-1G
102839-974EA 806.15 USD
102839-974
tert-Butyl-6-oxo-1,6-dihydropyridazine-3-carboxylate ≥97%
tert-Butyl-6-oxo-1,6-dihydropyridazine-3-carboxylate

Hazard: Irritant

Formula: C₉H₁₂N₂O₃
MW: 196.21 g/mol
MDL Number: MFCD19706069
CAS Number: 1355334-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR