tert-Butyl-6-oxo-1,4,5,6-tetrahydropyridazine-3-carboxylate ≥97%
Supplier: Matrix Scientific
083511-1G
102839-972EA
592.31
USD
102839-972
tert-Butyl-6-oxo-1,4,5,6-tetrahydropyridazine-3-carboxylate ≥97%
tert-Butyl-6-oxo-1,4,5,6-tetrahydropyridazine-3-carboxylate
Hazard: Irritant
Formula:
C₉H₁₄N₂O₃ MW: 198.22 g/mol Melting Pt: 146…148 °C |
MDL Number:
MFCD18089233 CAS Number: 434331-85-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...