Methyl-2-amino-5-(trifluoromethyl)-1,3-thiazole-4-carboxylate ≥97%

Supplier: Matrix Scientific

083468-1G
102839-860EA 1187.69 USD
102839-860
Methyl-2-amino-5-(trifluoromethyl)-1,3-thiazole-4-carboxylate ≥97%
Methyl-2-amino-5-(trifluoromethyl)-1,3-thiazole-4-carboxylate

Hazard: Irritant

Formula: C₆H₅F₃N₂O₂S
MDL Number: MFCD09991785
CAS Number: 1086375-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR