Methyl [5-(chloroacetyl)-1-methyl-1H-pyrrol-2-yl]acetate ≥97%
Supplier: Matrix Scientific
083459-1G
102839-838EA
750.77
USD
102839-838
Methyl [5-(chloroacetyl)-1-methyl-1H-pyrrol-2-yl]acetate ≥97%
Methyl [5-(chloroacetyl)-1-methyl-1H-pyrrol-2-yl]acetate
Hazard: Irritant
Formula:
C₁₀H₁₂ClNO₃ |
MDL Number:
MFCD09842625 CAS Number: 617721-31-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...