Ethyl-4-methyl-2-(1H-pyrrol-1-yl)thiophene-3-carboxylate ≥97%

Supplier: Matrix Scientific

083446-1G
102839-804EA 863.08 USD
102839-804
Ethyl-4-methyl-2-(1H-pyrrol-1-yl)thiophene-3-carboxylate ≥97%
Ethyl-4-methyl-2-(1H-pyrrol-1-yl)thiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₃NO₂S
MW: 235.31 g/mol
MDL Number: MFCD06808670
CAS Number: 874832-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR