Methyl-3-{2-[4-(tert-butoxycarbonyl)piperazin-1-yl]ethoxy}thiophene-2-carboxylate ≥97%

Supplier: Matrix Scientific

083436-1G
102839-776EA 636.92 USD
102839-776
Methyl-3-{2-[4-(tert-butoxycarbonyl)piperazin-1-yl]ethoxy}thiophene-2-carboxylate ≥97%
Methyl-3-{2-[4-(tert-butoxycarbonyl)piperazin-1-yl]ethoxy}thiophene-2-carboxylate

Hazard: Irritant

Formula: C₁₇H₂₆N₂O₅S
MDL Number: MFCD11100103
CAS Number: 1215799-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR