Methyl-4-piperidin-1-ylpyridine-2-carboxylate ≥97%
Supplier: Matrix Scientific
083425-1G
102839-740EA
750.77
USD
102839-740
Methyl-4-piperidin-1-ylpyridine-2-carboxylate ≥97%
Methyl-4-piperidin-1-ylpyridine-2-carboxylate
Hazard: Irritant
Formula:
C₁₂H₁₆N₂O₂ |
MDL Number:
MFCD19706064 CAS Number: 1355334-43-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...