Ethyl-3-amino-5-bromoisonicotinate ≥97%

Supplier: Matrix Scientific

083419-1G
102839-728EA 766.15 USD
102839-728
Ethyl-3-amino-5-bromoisonicotinate ≥97%
Ethyl-3-amino-5-bromoisonicotinate

Hazard: Irritant

Formula: C₈H₉BrN₂O₂
MDL Number: MFCD16622248
CAS Number: 1257535-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR