Ethyl difluoro(5-methylpyridin-2-yl)acetate ≥97%

Supplier: Matrix Scientific

083410-1G
102839-708EA 666.15 USD
102839-708
Ethyl difluoro(5-methylpyridin-2-yl)acetate ≥97%
Ethyl difluoro(5-methylpyridin-2-yl)acetate

Hazard: Irritant

Formula: C₁₀H₁₁F₂NO₂
MDL Number: MFCD12827881
CAS Number: 503627-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR