Methyl-2-phenoxyisonicotinate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 2-phenoxyisonicotinate

083403-1G
102839-690EA 636.92 USD
102839-690
Methyl-2-phenoxyisonicotinate ≥97%
Methyl-2-phenoxyisonicotinate

Hazard: Irritant

Formula: C₁₃H₁₁NO₃
MW: 229.24 g/mol
MDL Number: MFCD11505025
CAS Number: 1217114-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR