Methyl-5-bromo-2-ethoxynicotinate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 5-Bromo-2-ethoxynicotinate

083399-1G
102839-674EA 383.08 USD
102839-674
Methyl-5-bromo-2-ethoxynicotinate ≥97%
Methyl-5-bromo-2-ethoxynicotinate

Hazard: Irritant

Formula: C₉H₁₀BrNO₃
MW: 260.09 g/mol
MDL Number: MFCD09842613
CAS Number: 1031927-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR