Methyl-4-chloropyridine-2-carboxylate 1-oxide ≥97%

Supplier: Matrix Scientific

083382-1G
102839-628EA 1118.46 USD
102839-628
Methyl-4-chloropyridine-2-carboxylate 1-oxide ≥97%
Methyl-4-chloropyridine-2-carboxylate 1-oxide

Hazard: Irritant

Formula: C₇H₆ClNO₃
MDL Number: MFCD17015049
CAS Number: 90048-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR